Learn More
Etofenamate, Mikromol™
Description
Etofenamate
Specifications
Specifications
| Chemical Name or Material | Etofenamate |
| CAS | 30544-47-9 |
| Analyte or Component Names | Etofenamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 F3 N O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Etofenamate, 2-(2-Hydroxyethoxy)ethyl 2-[[3-(trifluoromethyl)phenyl]amino]benzoate, Benzoic acid, 2-[[3-(trifluoromethyl)phenyl]amino]-, 2-(2-hydroxyethoxy)ethyl ester, 2-(2-Hydroxyethoxy)ethyl flufenamate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid 2-(2-hydroxyethoxy)ethyl ester, B 577, Bay-d 1107, Bayrogel, Glasel, Reumon, Rheumon, Rheumon gel, TV 485, Traumon Gel, WHR 5020, Anthranilic acid, N-(alpha,alpha,alpha-trifluoro-m-tolyl)-, 2-(2-hydroxyethoxy)ethyl ester (8CI) |
| InChI Formula | InChI=1S/C18H18F3NO4/c19-18(20,21)13-4-3-5-14(12-13)22-16-7-2-1-6-15(16)17(24)26-11-10-25-9-8-23/h1-7,12,22-23H,8-11H2 |
| SMILES | OCCOCCOC(C1=CC=CC=C1NC2=CC=CC(C(F)(F)F)=C2)=O |
| IUPAC Name | 2-(2-hydroxyethoxy)ethyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |