Learn More
ETNPPL Isoform 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5113882
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: KRVLLSADGP HRNVLKIKPP MCFTEEDAKF MVDQLDRILT VLEEAMGTKT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
Catalyzes the pyridoxal-phosphate-dependent breakdown of phosphoethanolamine, converting it to ammonia, inorganic phosphate and acetaldehyde.Specifications
ETNPPL Isoform 1 | |
Polyclonal | |
Unconjugated | |
ETNPPL | |
1300019H02Rik; Agxt2l1; AI195447; alanine--glyoxylate aminotransferase 2-like 1; alanine-glyoxylate aminotransferase 2-like 1; ethanolamine phosphate phospholyase; ethanolamine-phosphate phospho-lyase; ETNPPL | |
Rabbit | |
Affinity chromatography | |
RUO | |
64850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TBG4 | |
ETNPPL | |
Synthetic peptide directed towards the middle region of human AGXT2L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |