missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETNPPL Isoform 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5104423
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total AGXT2L1.
Catalyzes the pyridoxal-phosphate-dependent breakdown of phosphoethanolamine, converting it to ammonia, inorganic phosphate and acetaldehyde.Specifications
ETNPPL Isoform 1 | |
Polyclonal | |
Unconjugated | |
ETNPPL | |
1300019H02Rik; Agxt2l1; AI195447; alanine--glyoxylate aminotransferase 2-like 1; alanine-glyoxylate aminotransferase 2-like 1; ethanolamine phosphate phospholyase; ethanolamine-phosphate phospho-lyase; ETNPPL | |
Rabbit | |
Affinity chromatography | |
RUO | |
64850, 687071, 71760 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BWU8, Q8TBG4 | |
ETNPPL | |
A synthesized peptide derived from human ETNPPL(Accession Q8TBG4), corresponding to amino acid residues P271-L321. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |