missing translation for 'onlineSavingsMsg'
Learn More

Etifoxine hydrochloride, MedChemExpress

Product Code. 16457126
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457126 100 mg 100mg
16459627 5 mg 5mg
16453127 50 mg 50mg
16407088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16457126 Supplier Medchem Express Supplier No. HY16579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents.

Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents.

Specifications

Chemical Name or Material Etifoxine hydrochloride
Physical Form Solid
CAS 56776-32-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H18Cl2N2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym HOE 36-801 hydrochloride
Solubility Information DMSO : 50 mg/mL (148.26 mM; Need ultrasonic) ∣H2O : 5 mg/mL (14.83 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(C(C3=CC=CC=C3)(C)OC(NCC)=N2)=C1.Cl
Molecular Weight (g/mol) 337.24
Formula Weight 337.24
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.