Learn More
Ethynodiol Diacetate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ethynodiol Diacetate |
| CAS | 297-76-7 |
| Analyte or Component Names | Ethynodiol Diacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 8080CB, Cervicundin, Continuin, Ethinodiol diacetate, Ethynodiol acetate, Ethynodiol diacetate, Femulen, Luteonorm, Luto-Metrodiol, Metrodiol, Metrodiol, diacetate, SC 11800, 19-Norpregn-4-en-20-yne-3,17-diol, 3,17-diacetate, (3beta,17alpha)-, 19-Nor-17alpha-pregn-4-en-20-yne-3beta,17-diol, diacetate (7CI,8CI), 19-Norpregn-4-en-20-yne-3,17-diol, diacetate, (3beta,17alpha)- (9CI), 17-Ethynyl-19-norandrost-4-ene-3beta,17beta-diol diacetate, 17alpha-Ethynyl-4-estren-3beta,17-diol diacetate, 17alpha-Ethynyl-4-estrene-3beta,17beta-diacetate, 17alpha-Ethynyl-4-estrene-3beta,17beta-diol diacetate, 3beta,17beta-Diacetoxy-19-nor-17alpha-pregn-4-en-20-yne, beta-Ethynodiol diacetate |
| InChI Formula | InChI=1S/C24H32O4/c1-5-24(28-16(3)26)13-11-22-21-8-6-17-14-18(27-15(2)25)7-9-19(17)20(21)10-12-23(22,24)4/h1,14,18-22H,6-13H2,2-4H3/t18-,19-,20+,21+,22-,23-,24-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)O[C@H]1CC[C@@H]2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(OC(=O)C)C#C)[C@@H]3CCC2=C1 |
| Show More |