Learn More
Ethylenedinitrilotetraacetic Acid Dipotassium Salt Dihydrate, Supelco™
Specifications
Specifications
| Melting Point | 272°C |
| CAS | 25102-12-9 |
| Quantity | 1 kg |
| MDL Number | MFCD00150036 |
| Synonym | Ethylenedinitrilotetraacetic acid dipotassium salt dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt, Titriplex™ Potassium salt, Ethylenedinitrilotetraacetic acid dipotassium salt dihydrate |
| Molecular Weight (g/mol) | 404.45 |
| Grade | Research |
| Chemical Name or Material | Ethylenedinitrilotetraacetic Acid Dipotassium Salt Dihydrate |