Learn More
Ethylenediamine, Sigma-Aldrich™
Product Code. 15868470
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
50 L
Unit Size:
1 L
100mL
2.5 L
50 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15868470 50 L 50 L
15838470 100 mL 100mL
15848470 500 mL 500mL
15806280 1 L 1 L
15858470 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15868470 Supplier Sigma Aldrich Supplier No. 8009479051
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-15-3
Specifications
Melting Point 8.5°C (lit.)
Density 0.899 g/mL at 25°C
Boiling Point 118°C
Molecular Formula NH2CH2CH2NH2
MDL Number MFCD00008204
Quantity 50 L
Synonym 1,2-Diaminoethane, 1,2-Ethanediamine, 1,2-Diaminoethane, 1,2-Diaminoethane
Molecular Weight (g/mol) 60.1
Grade Research