missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Spectrophotometric grade, 99+%

Product Code. p-4408353
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11485270

Brand: Thermo Scientific Alfa Aesar 044529.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

For use in spectrophotometryEthylene glycol is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point °C to 13°C
Density 1.113 g/mL
Boiling Point 196°C to 198°C
Flash Point 119°C (246°F)
Assay Percent Range >99%
Refractive Index 1.431
Quantity 1 L
Beilstein 505945
Sensitivity Hygroscopic
Solubility Information Miscible with water,toluene,tetrahydrofuran,alcohol,chloroform and other organic solvents.
Formula Weight 62.07
Grade Spectrophotometric
Physical Form Liquid
Chemical Name or Material Ethylene glycol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.