Learn More
Ethylene Glycol Dimethyl Ether, Sigma-Aldrich™
Description
Ethylene glycol dimethyl ether, also known as dimethoxyethane, has gained significant attention in recent years due to its favorable characteristics, including a high energy density and cetane number. It possesses excellent solubility properties and is used as an environmentally friendly solvent and effective etherification agent in industries such as cosmetics, perfumes, and pharmaceuticals. Notably, it finds extensive application in batteries and electrolytes due to its desirable properties.
Specifications
Specifications
| Melting Point | −58°C (Lit.) |
| CAS | 110-71-4 |
| Density | 0.867 g/mL at 25°C |
| Quantity | 2.5 L |
| Molecular Formula | CH3OCH2CH2OCH3 |
| MDL Number | MFCD00008502 |
| Synonym | 1,2-Dimethoxyethane, mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme, 1,2-Dimethoxyethane, mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme |
| Boiling Point | 85°C |
| Molecular Weight (g/mol) | 90.12 |
| Grade | Research |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.