missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol-Bis (Succinic Acid N-Hydroxysuccinimide Ester), MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Cross Linking Reagent
Specifications
Specifications
| CAS | 70539-42-3 |
| Color | White |
| Molecular Formula | C18H20N2O12 |
| MDL Number | MFCD00049057 |
| Quantity | 250 mg |
| Synonym | ethylene glycol-bis succinic acid n-hydroxysuccinimide ester, ethylene glycol bis succinimidyl succinate, bicl110, 1,2-ethanediyl bis 4-2,5-dioxo-1-pyrrolidinyl oxy-4-oxobutanoate, ethylene glycol-bis succinimidyl succinate, bis 2,5-dioxopyrrolidin-1-yl o,o'-ethane-1,2-diyl disuccinate, 2,5-dioxopyrrolidin-1-yl 1-2-4-2,5-dioxopyrrolidin-1-yl oxy-4-oxobutanoyl oxy ethyl butanedioate, egs, egbss, ethylene glycolylbis succinimidyl succinate |
| InChI Key | QLHLYJHNOCILIT-UHFFFAOYSA-N |
| SMILES | O=C(CCC(=O)ON1C(=O)CCC1=O)OCCOC(=O)CCC(=O)ON1C(=O)CCC1=O |
| IUPAC Name | 2,5-dioxopyrrolidin-1-yl 1-[2-({4-[(2,5-dioxopyrrolidin-1-yl)oxy]-4-oxobutanoyl}oxy)ethyl] butanedioate |
| Molecular Weight (g/mol) | 456.36 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction