missing translation for 'onlineSavingsMsg'
Learn More

Ethyl palmitate, 97%

Product Code. 11439170
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11439170

Brand: Thermo Scientific Alfa Aesar A15694.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-97-7 | C18H36O2 | 284.48 g/mol

Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

Solubility
Miscible with ethanol, dimethyl sulfoxide, chloroform and ethyl acetate. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-97-7
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00008996
InChI Key XIRNKXNNONJFQO-UHFFFAOYSA-N
Synonym ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
PubChem CID 12366
ChEBI CHEBI:84932
IUPAC Name ethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 22°C to 26°C
Density 0.857
Boiling Point 192°C to 193°C (10 mmHg)
Flash Point 178°C (352°F)
Refractive Index 1.4395
Quantity 100 g
Beilstein 1782663
Solubility Information Miscible with ethanol,dimethyl sulfoxide,chloroform and ethyl acetate. Immiscible with water.
Formula Weight 284.48
Percent Purity 97%
Chemical Name or Material Ethyl palmitate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.