Learn More
Ethyl nicotinate, 99%
Product Code. 11322066
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. 11322066
Brand: Thermo Scientific Alfa Aesar L03215.36
Request Bulk or Custom Format
CAS: 614-18-6 | C8H9NO2 | 151.165 g/mol
Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.
Solubility
Miscible with water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 614-18-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006389
InChI Key XBLVHTDFJBKJLG-UHFFFAOYSA-N
Synonym ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan
PubChem CID 69188
IUPAC Name ethyl pyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=CC=C1
Specifications
Melting Point 8°C to 9°C
Density 1.107
Boiling Point 223°C to 224°C
Flash Point 93°C (199°F)
Odor Characteristic
Refractive Index 1.503
Quantity 500 g
Beilstein 122937
Solubility Information Miscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Ethyl nicotinate