missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Ketone, Supelco™

Product Code. 15874740
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
190 L
Unit Size:
1 L
190 L
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874740 190 L 190 L
15834740 1 L 1 L
15844740 2.5 L 2.5 L
15864740 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15874740 Supplier Supelco Supplier No. 1060149190
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Butanone EMPLURA™

EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3

Specifications

Melting Point -86°C
Density 0.805 g/cm3 at 20°C
Boiling Point 79.6°C/1013 hPa
MDL Number MFCD00011648
Quantity 190 L
Synonym Ethyl methyl ketone, 2-Butanone, Methyl ethyl ketone, MEK, Ethyl methyl ketone
Molecular Weight (g/mol) 72.11
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.