missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Ketone, Supelco™

Product Code. 15825390
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15825390 1 L 1 L
15807770 2.5 L 2.5 L
15817770 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15825390 Supplier Supelco Supplier No. 1097081000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,Reag. Ph Eur

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3

Specifications

Melting Point -86°C
Density 0.805 g/cm3 at 20°C
Boiling Point 79.6°C/1013 hPa
MDL Number MFCD00011648
Quantity 1 L
Synonym Ethyl methyl ketone, 2-Butanone, Methyl ethyl ketone, MEK, Ethyl methyl ketone
Molecular Weight (g/mol) 72.11
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.