Learn More
Ethyl Methyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol™
Description
Ethyl Methyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate
Specifications
Specifications
| Chemical Name or Material | Ethyl Methyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate |
| CAS | 103890-81-9 |
| Analyte or Component Names | Ethyl Methyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H31 N O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl Methyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, Lacidipine Imp. A (BP), Lacidipine Impurity A |
| InChI Formula | InChI=1S/C25H31NO6/c1-8-31-24(29)21-16(3)26-15(2)20(23(28)30-7)22(21)18-12-10-9-11-17(18)13-14-19(27)32-25(4,5)6/h9-14,22,26H,8H2,1-7H3/b14-13+ |
| SMILES | CCOC(=O)C1=C(C)NC(=C(C1c2ccccc2\C=C\C(=O)OC(C)(C)C)C(=O)OC)C |
| IUPAC Name | 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate |
| Show More |