Learn More
Ethyl Laurate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 106-33-2 |
| Melting Point | -2°C |
| Density | 0.86 g/cm3 at 20°C |
| Boiling Point | 272°C/1013 hPa |
| Molecular Formula | CH3(CH2)10COOC2H5 |
| MDL Number | MFCD00015065 |
| Quantity | 250 mL |
| Synonym | Ethyl laurate, Ethyl dodecanoate, Lauric acid ethyl ester, Ethyl laurate |
| Molecular Weight (g/mol) | 228.37 |
| Grade | Research |