missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen glutarate, 97%, Thermo Scientific Chemicals

Product Code. 11396716
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396716 25 g 25g
11386716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396716 Supplier Thermo Scientific Alfa Aesar Supplier No. L03941.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-62-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00014379
InChI Key MYMNBFURSYZQBR-UHFFFAOYSA-N
Synonym ethyl hydrogen glutarate, glutaric acid monoethyl ester, 5-ethoxy-5-oxo-pentanoic acid, ethyl glutarate,mono, acmc-20akx9, ksc493i1d, pentanedioic acid 1-ethyl ester, pentanedioic acid monoethyl ester, 5-ethoxy-5-oxidanylidene-pentanoic acid, glutaric acid monoethyl ester; monoethyl glutarate
PubChem CID 66114
IUPAC Name 5-ethoxy-5-oxopentanoic acid
SMILES CCOC(=O)CCCC(=O)O

Specifications

Boiling Point 118°C to 120°C (1.5 mmHg)
Refractive Index 1.4365
Quantity 25 g
Beilstein 1766397
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Ethyl hydrogen glutarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.