missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dibromofluoroacetate, 97%

Product Code. p-7046830
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11399195

Brand: Thermo Scientific Alfa Aesar L16491.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-53-7 | C4H5Br2FO2 | 263.89 g/mol

Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-53-7
Molecular Formula C4H5Br2FO2
Molecular Weight (g/mol) 263.89
MDL Number MFCD00042068
InChI Key COYRDMUQLSIAEX-UHFFFAOYSA-N
Synonym ethyl dibromofluoroacetate, dibromofluoroacetic acid ethyl ester, ethyldibromofluoroacetate, 2,2-dibromo-1-ethoxy-2-fluoroethan-1-one, acmc-1aqw2, ethyl 2,2-dibromo-2-fluoracetate, ethyl 2,2-dibromo-2-fluoro-acetate
PubChem CID 2737168
IUPAC Name ethyl 2,2-dibromo-2-fluoroacetate
SMILES CCOC(=O)C(F)(Br)Br

Specifications

Density 1.894
Boiling Point 82°C (33 mmHg)
Flash Point >110°C (230°F)
Linear Formula Br2CFCO2CH2CH3
Refractive Index 1.4635
Quantity 5 g
UN Number UN2922
Beilstein 1762048
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 263.89
Percent Purity 97%
Chemical Name or Material Ethyl dibromofluoroacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.