Learn More
Ethyl-d5 Carbamate, CDN
Specifications
Specifications
| Chemical Name or Material | Urethane-d5 |
| CAS | 73962-07-9 |
| Molecular Formula | C3 2H5 H2 N O2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OC(=O)N |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethyl carbamate |
| Molecular Weight (g/mol) | 94.12 |
| Formula Weight | 94.0791 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |