Learn More
Ethyl cyclopropanecarboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 4606-07-9 |
| Molecular Formula | C6H10O2 |
| MDL Number | MFCD00001282 |
| Quantity | 5 g |
| SMILES | O=C(C1CC1)OCC |
| Molecular Weight (g/mol) | 114.14 |
| Percent Purity | 97% |
| Chemical Name or Material | Ethyl cyclopropanecarboxylate |