Learn More
Ethyl benzimidate hydrochloride, 97%, BLD Pharm™
Product Code. 31005250
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31005250 10 g 10g
30990525 500 g 500g
30992812 100 g 100g
30990208 25 g 25g
30995526 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31005250 Supplier BLD Pharm Supplier No. BD12926310g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 5333-86-8
Molecular Formula C9H12ClNO
MDL Number MFCD00043246
Quantity 10 g
SMILES N=C(OCC)C1=CC=CC=C1.[H]Cl
Molecular Weight (g/mol) 185.65
Percent Purity 97%
Chemical Name or Material Ethyl benzimidate hydrochloride