missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol

Product Code. 13127603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13127603 2.5 L Glass Bottle 2.5 L
10338360 500 mL Glass Bottle 500mL
10481071 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 13127603 Supplier Thermo Scientific Acros Supplier No. 224090025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Absorbance 0.01 max. at 300nm, 0.04 max. at 250nm, 0.65 max. at 210nm, 1 max. at 205nm
Density 1.59
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.0005% max.
Linear Formula C2H5OH
Vapor Pressure Vapor Pressure: 59mbar at 20°C
Water 0.1% Max. (Coulometric)
Merck Index 14, 3761
Formula Weight 46.06
Percent Purity 4.0 to 6.0% (2-propanol)
Grade Spectroscopy
Chemical Name or Material Ethanol
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.