Learn More
Ethyl 8-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific™
Product Code. 15476038
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476038 1 g 1g
15415176 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15476038 Supplier Thermo Scientific Alfa Aesar Supplier No. H59657.03
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 206258-97-1
Molecular Formula C12H9BrClNO2
Molecular Weight (g/mol) 314.563
MDL Number MFCD00173385
InChI Key MLHAFVCMBUQWKC-UHFFFAOYSA-N
Synonym 8-bromo-4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl8-bromo-4-chloroquinoline-3-carboxylate, ethyl 4-chloro-8-bromo-3-quinolinecarboxylate, 4-chloro-8-bromoquinoline-3-carboxylic acid ethyl ester, 8-bromo-4-chloro-3-quinolinecarboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-bromo-4-chloro-, ethyl ester, 8-bromo-4-chloro-quinoline-3-carboxylic acid ethyl ester
PubChem CID 3418677
IUPAC Name ethyl 8-bromo-4-chloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C(=C1Cl)C=CC=C2Br
Specifications
Quantity 1 g
Formula Weight 314.57
Percent Purity 97%
Chemical Name or Material Ethyl 8-bromo-4-chloroquinoline-3-carboxylate