Learn More
Ethyl 7-azaindole-4-carboxylate, 97%, Thermo Scientific Chemicals
Product Code. 15457187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457187 1 g 1g
15467187 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15457187 Supplier Thermo Scientific Alfa Aesar Supplier No. H34250.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1261588-72-0
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD15071133
InChI Key CKCBQXQNAUHBDX-UHFFFAOYSA-N
PubChem CID 56773892
IUPAC Name ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate
SMILES CCOC(=O)C1=C2C=CNC2=NC=C1
Specifications
Melting Point 127°C to 129°C
Quantity 1 g
Synonym ethyl 1h-pyrrolo 2,3-b pyridine-4-carboxylate, c9e, ethylpyrrolobpyridinecarboxylate, ethyl 7-azaindole-4-carboxylate, ethyl1h-pyrrolo 2,3-b pyridine-4-carboxylate, 1h-pyrrolo 2,3-b pyridine-4-carboxylic acid, ethyl ester
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material Ethyl 7-azaindole-4-carboxylate