Learn More
Ethyl 5,6-Dimethylnicotinate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 5,6-Dimethylnicotinate |
| CAS | 77629-53-9 |
| Molecular Formula | C10 H13 N O2 |
| Quantity | 250 mg |
| Synonym | 3-Pyridinecarboxylic acid, 5,6-dimethyl-, ethyl ester, 5,6-Dimethyl-3-pyridinecarboxylic acid ethyl ester, 5,6-Dimethylpyridine-3-carboxylic acid ethyl ester, Ethyl 5,6-dimethyl-3-pyridinecarboxylate, Ethyl 5,6-dimethylnicotinate |
| InChI Formula | InChI=1S/C10H13NO2/c1-4-13-10(12)9-5-7(2)8(3)11-6-9/h5-6H,4H2,1-3H3 |
| SMILES | CCOC(=O)c1cnc(C)c(C)c1 |
| IUPAC Name | ethyl 5,6-dimethylpyridine-3-carboxylate |
| Molecular Weight (g/mol) | 179.22 |
| Formula Weight | 179.0946 |