Learn More
Ethyl (4-Aminophenyl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl (4-Aminophenyl)carbamate |
| CAS | 57399-97-0 |
| Molecular Formula | C9 H12 N2 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H12N2O2/c1-2-13-9(12)11-8-5-3-7(10)4-6-8/h3-6H,2,10H2,1H3,(H,11,12) |
| SMILES | CCOC(=O)Nc1ccc(N)cc1 |
| IUPAC Name | ethyl N-(4-aminophenyl)carbamate |
| Molecular Weight (g/mol) | 180.204 |
| Formula Weight | 180.09 |