missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-nitrocinnamate, 98%, Thermo Scientific™

Product Code. 11439883
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439883 5 g 5g
11449883 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439883 Supplier Thermo Scientific Alfa Aesar Supplier No. A12995.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5396-71-4
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD00017027
InChI Key QZEPRSLOWNHADS-VOTSOKGWSA-N
Synonym ethyl 3-nitrocinnamate, ethyl 3-3-nitrophenyl acrylate, ethyl 2e-3-3-nitrophenyl prop-2-enoate, 2-propenoic acid, 3-3-nitrophenyl-, ethyl ester, ethyltrans-3-nitrocinnamate, e-ethyl 3-3-nitrophenyl acrylate, ethyl-3-nitrocinnamate, ethyl 3-nitrocinnamicacid, ethyl trans-3-nitrocinnamate
PubChem CID 5354248
IUPAC Name ethyl (E)-3-(3-nitrophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 74°C to 76°C
Quantity 5 g
Beilstein 2618102
Formula Weight 221.21
Percent Purity 98%
Chemical Name or Material Ethyl 3-nitrocinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.