Learn More
Ethyl 3-hydroxybenzoate, 99%
Product Code. 11458256
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11458256 25 g 25g
11468256 100 g 100g
11478256 500 g 500g
3 options
Product Code. 11458256
Brand: Thermo Scientific Alfa Aesar A15343.14
Request Bulk or Custom Format
CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7781-98-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002296
InChI Key MWSMNBYIEBRXAL-UHFFFAOYSA-N
Synonym ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate
PubChem CID 24522
IUPAC Name ethyl 3-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)O
Specifications
Melting Point 72°C to 76°C
Boiling Point 187°C to 188°C (31 mmHg)
Quantity 25 g
Beilstein 2208566
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 3-hydroxybenzoate