missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-hexynoate, 98+%, Thermo Scientific™

Catalog Number 11321857
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
16205-90-6
C8H12O2
140.182
MFCD00015257
ZFMNCZDBAHELNX-UHFFFAOYSA-N
ethyl 2-hexynoate, 2-hexynoic acid, ethyl ester, 2-hexynoicacid,ethyl ester, acmc-20al13, 2-hexynoic acid ethyl ester, 2-hexynoicacid, ethyl ester, hex-2-ynoic acid ethyl ester, ethyl 2-hexynoate,
85329
ethyl hex-2-ynoate
CCCC#CC(=O)OCC
This item is not returnable. View return policy
16205-90-6
C8H12O2
140.182
MFCD00015257
ZFMNCZDBAHELNX-UHFFFAOYSA-N
ethyl 2-hexynoate, 2-hexynoic acid, ethyl ester, 2-hexynoicacid,ethyl ester, acmc-20al13, 2-hexynoic acid ethyl ester, 2-hexynoicacid, ethyl ester, hex-2-ynoic acid ethyl ester, ethyl 2-hexynoate,
85329
ethyl hex-2-ynoate
CCCC#CC(=O)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 87°C to 88°C (14 mmHg)
Linear Formula CH3CH2CH2C≡CCO2CH2CH3
Refractive Index 1.442
Quantity 2 g
Beilstein 1702264
Solubility Information Insoluble in water.
Formula Weight 140.18
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-hexynoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'