Learn More
Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate |
| CAS | 161797-99-5 |
| Molecular Formula | C13H13NO3S |
| Quantity | 250 mg |
| Synonym | 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylic Acid Ethyl Ester, Ethyl 2-(4-Hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate |
| InChI Formula | InChI=1S/C13H13NO3S/c1-3-17-13(16)11-8(2)14-12(18-11)9-4-6-10(15)7-5-9/h4-7,15H,3H2,1-2H3 |
| SMILES | OC1=CC=C(C2=NC(C)=C(C(OCC)=O)S2)C=C1 |
| IUPAC Name | ethyl 2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate |
| Molecular Weight (g/mol) | 263.31 |
| Formula Weight | 263.06 |