Learn More
Ethyl 2,4-difluorobenzoate, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Boiling Point | 65°C to 67°C (15 mmHg) |
| Quantity | 5 g |
| Formula Weight | 186.16 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 2,4-difluorobenzoate |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.