Learn More
Ethyl 2,4-difluorobenzoate, 98%, Thermo Scientific™
Product Code. 11454574
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11454574
Brand: Thermo Scientific Alfa Aesar A13177.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108928-00-3
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.16
MDL Number MFCD00153149
InChI Key OPQFYGPAOVCNEQ-UHFFFAOYSA-N
Synonym 2,4-difluorobenzoic acid ethyl ester, 2,4-difluoro-benzoic acid ethyl ester, pubchem3467, acmc-209w2m, ethyl-2,4-difluorobenzoate, ethyl 2,4-di-fluorobenzoate, rarechem al bi 0213, ethyl2,4-difluorobenzoate, 2,4 ethyl-difluorobenzoate, 2,4-difluorobenzoic acid, ethyl ester
PubChem CID 2737170
IUPAC Name ethyl 2,4-difluorobenzoate
SMILES CCOC(=O)C1=CC=C(F)C=C1F
Specifications
Boiling Point 65°C to 67°C (15 mmHg)
Quantity 1 g
Formula Weight 186.16
Percent Purity 98%
Chemical Name or Material Ethyl 2,4-difluorobenzoate