Learn More
Ethyl 2-(3-N-phthalimidopropyl)acetoacetate, 95%, Thermo Scientific™
Catalog Number 11333656
Click to view available options
Quantity:
5 g
25 g
55747-45-0
C17H19NO5
317.341
MFCD00399474
XDGCOSMBEZEXSN-UHFFFAOYSA-N
ethyl 2-3-n-phthalimidopropyl acetoacetate, ethyl 2-acetyl-5-1,3-dioxoisoindolin-2-yl pentanoate, ethyl 2-acetyl-5-1,3-dioxoisoindol-2-yl pentanoate, ethyl 2-3-phthalimidopropyl acetoacetate, ethyl alpha-acetyl-delta-phthalimidovalerate, 2-acetyl-5-phthalimidylvaleric acid ethyl ester, ethyl 2-acetyl-5-1,3-dioxobenzo c azolidin-2-yl pentanoate
4071008
ethyl 2-acetyl-5-(1,3-dioxoisoindol-2-yl)pentanoate
CCOC(=O)C(CCCN1C(=O)C2=CC=CC=C2C1=O)C(=O)C
55747-45-0
C17H19NO5
317.341
MFCD00399474
XDGCOSMBEZEXSN-UHFFFAOYSA-N
ethyl 2-3-n-phthalimidopropyl acetoacetate, ethyl 2-acetyl-5-1,3-dioxoisoindolin-2-yl pentanoate, ethyl 2-acetyl-5-1,3-dioxoisoindol-2-yl pentanoate, ethyl 2-3-phthalimidopropyl acetoacetate, ethyl alpha-acetyl-delta-phthalimidovalerate, 2-acetyl-5-phthalimidylvaleric acid ethyl ester, ethyl 2-acetyl-5-1,3-dioxobenzo c azolidin-2-yl pentanoate
4071008
ethyl 2-acetyl-5-(1,3-dioxoisoindol-2-yl)pentanoate
CCOC(=O)C(CCCN1C(=O)C2=CC=CC=C2C1=O)C(=O)C
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 62°C to 64°C
Quantity 5 g
Formula Weight 317.34
Percent Purity 95%
Chemical Name or Material Ethyl 2-(3-N-phthalimidopropyl)acetoacetate