Learn More
Ethyl 2,3-dibromobutanoate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 609-11-0 |
| Molecular Formula | C6H10Br2O2 |
| MDL Number | MFCD00017863 |
| Quantity | 5 g |
| SMILES | CC(Br)C(Br)C(OCC)=O |
| Molecular Weight (g/mol) | 273.95 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethyl 2,3-dibromobutanoate |