Learn More
Ethyl 1-Methylimidazole-2-carboxylate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Ethyl 1-Methylimidazole-2-carboxylate |
| CAS | 30148-21-1 |
| Molecular Formula | C7 H10 N2 O2 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C7H10N2O2/c1-3-11-7(10)6-8-4-5-9(6)2/h4-5H,3H2,1-2H3 |
| SMILES | CCOC(=O)c1nccn1C |
| IUPAC Name | ethyl 1-methylimidazole-2-carboxylate |
| Molecular Weight (g/mol) | 154.17 |
| Formula Weight | 154.0742 |