missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate, 95%

Product Code. p-7041484
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487236 250 mg 250mg
15467236 1 g 1g
15477236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487236 Supplier Thermo Scientific Alfa Aesar Supplier No. H62277.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3697-68-5 | C7H12O3 | 144.17 g/mol

Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.

Notes
Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3697-68-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD09759206
InChI Key PAILVKQSHRJIPE-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclopropanecarboxylate, ethyl 1-hydroxymethyl-cyclopropanecarboxylate, 1-hydroxymethyl-cyclopropanecarboxylic acid ethyl ester, ethyl 1-hydroxymethyl cyclopropane-1-carboxylate, ethyl 1-hydroxymethylcyclopropanecarboxylate, ethyl1-hydroxymethyl-cyclopropanecarboxylate, ethyl 1-hydroxymethyl-1-cyclopropanecarboxylate, 1-hydroxymethyl-cyclopropanecarboxylic acid ethyl, 1-hydroxymethylcyclopropanecarboxylic acid ethyl ester, 1-hydroxymethyl cyclopropane carboxylic acid ethyl ester
PubChem CID 12450336
IUPAC Name ethyl 1-(hydroxymethyl)cyclopropane-1-carboxylate
SMILES CCOC(=O)C1(CO)CC1

Specifications

Quantity 250 mg
Formula Weight 144.16
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.