Learn More
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate, 95%
Product Code. 15467236
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467236 1 g 1g
15487236 250 mg 250mg
15477236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15467236 Supplier Thermo Scientific Alfa Aesar Supplier No. H62277.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
CAS: 3697-68-5 | C7H12O3 | 144.17 g/mol
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
Notes
Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3697-68-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD09759206
InChI Key PAILVKQSHRJIPE-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclopropanecarboxylate, ethyl 1-hydroxymethyl-cyclopropanecarboxylate, 1-hydroxymethyl-cyclopropanecarboxylic acid ethyl ester, ethyl 1-hydroxymethyl cyclopropane-1-carboxylate, ethyl 1-hydroxymethylcyclopropanecarboxylate, ethyl1-hydroxymethyl-cyclopropanecarboxylate, ethyl 1-hydroxymethyl-1-cyclopropanecarboxylate, 1-hydroxymethyl-cyclopropanecarboxylic acid ethyl, 1-hydroxymethylcyclopropanecarboxylic acid ethyl ester, 1-hydroxymethyl cyclopropane carboxylic acid ethyl ester
PubChem CID 12450336
IUPAC Name ethyl 1-(hydroxymethyl)cyclopropane-1-carboxylate
SMILES CCOC(=O)C1(CO)CC1
Specifications
Quantity 1 g
Formula Weight 144.16
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate