Learn More
Ethanolamine, Sigma-Aldrich™
Product Code. 15876260
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
25 L
Unit Size:
1 L
100mL
2.5 L
25 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876260 1 L 1 L
15856260 100 mL 100mL
15866260 500 mL 500mL
15848430 2.5 L 2.5 L
15858430 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15876260 Supplier Sigma Aldrich Supplier No. 8008491000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-43-5
Specifications
Melting Point 10.5°C
Density 1.02 g/cm3 at 20°C
Boiling Point 171°C/1013 hPa
MDL Number MFCD00008183
Quantity 1 L
Synonym Ethanolamine, 2-Aminoethanol, Monoethanolamine, Ethanolamine
Molecular Weight (g/mol) 61.08
Grade Research