Learn More
Ethanol solution (Untaxed), Tested according to Ph.Helv., 70 % (v/v) in H2O, for license holders only, Solstice
Product Code. 15640630
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15640630 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15640630 Supplier Honeywell-Riedel-de Haen Supplier No. 028772.5L
  • ONLINE EXCLUSIVE PRICE
    1480.00 NOK
    1188.68 NOK / 2.5 L
    Save 291.32 NOK
    19% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Specifications
CAS 64-17-5
Quantity 2.5 L
Flash Point 22°C (71.6°F)
Molecular Formula C2H6O
Linear Formula C2H5OH
MDL Number MFCD00003568
UN Number UN1170
Beilstein 1718733
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity 0%
Chemical Name or Material Ethanol Solution, Tested According to Ph.Helv.