Learn More
Ethanol, Absolute, Pharmpur™, Scharlab
Product Code. 11374054
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity unitSize
11374054 1 L 1 L
1 options
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 11374054
Brand: Scharlab ET00061000
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Used in solvents, as a disinfectant, for pharmaceutical use, synthesis of organic products, and in perfumery.
  • Synonyms: Ethyl alcohol, Methylcarbinol, Spirit, Spirit of wine
  • Molecular Formula: C2H5OH
  • Molar Mass: 46.07g/mol
  • CAS: 64-17-5
  • EIN ECS No.: 200-578-6
  • Dielectric Constant: 24.3 at 25°C
  • Reducing Substances: Passes test
Reacts explosively with strong oxidizing agents including chromium trioxide, hydrogen peroxide and inorganic acids such as nitric acid. Can react violently with metals such as magnesium, sodium and potassium to produce hydrogen. May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.
Specifications
Melting Point -114.5°C
CAS 64-17-5
Appearance Clear and Colorless
Density 0.79g/cm3 
Boiling Point 78.3°C
Quantity 1 L
Flash Point 12°C
Identification Passes Test
Ignition Point 425°C
Physical Form Liquid
Residue after Evaporation 0.001% max
Molecular Formula C2H6O
Vapor Pressure 59hPa (at 20°C)
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Solubility Information Miscible in water at 20°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity 99.5% min (G.C., v/v), 99.2% min (G.C., w/w)
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol absolute, extra pure,Pharmpur, Ph Eur, BP, USP
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501-Dispose of contents/container to .