Learn More
Ethanol 70% in water (v/v) (Taxed), for domestic sale in Germany, France, Ireland and United Kingdom, Solstice
Product Code. 15668460
Change view
Click to view available options
Quantity:
26 mL
Packaging:
Poly Bottle
Unit Size:
26mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15668460 26 mL Poly Bottle 26mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15668460 Supplier Fluka Supplier No. 7229826ML
  • ONLINE EXCLUSIVE PRICE
    295.00 NOK
    236.82 NOK / 26mL
    Save 58.18 NOK
    19% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Specifications
CAS 64-17-5, 7732-18-5
CAS Min % ≥70.0000%, ≥30.0000%
CAS Max % <90.0000%, <50.0000%
Density 0.886 g/cm3
Quantity 26 mL
Flash Point 22°C
Packaging Poly Bottle
Assay Percent Range 69 to 71%
Molecular Formula C2H6O
Linear Formula (CH3CH2)2O
MDL Number MFCD00011646
UN Number UN1170
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 77°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Grade Performance
Assay Vol.
Chemical Name or Material Ethanol 70% in water (v/v) (Taxed), for domestic sale in Germany, France, Ireland and United Kingdom