Description
Outlet tubing for ÅKTA avant 25.
Specifications
Specifications
| Type | Tubing |
| Diameter (Metric) Outer | 1.5 mm |
| Diameter (Metric) Inner | 1 mm |
| Packaging Quantity | 1 Each |
| For Use With | For ÄKTA Avant 25 |
| Material | ETFE |