Learn More
Abnova™ ETF1 (Human) Recombinant Protein
Product Code. 16127814
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16127814 25 μg 25µg
16117814 10 μg 10µg
2 options
Product Code. 16127814
Brand: Abnova™ H00002107Q01.25ug
Human ETF1 partial ORF ( NP_004721.1, 338 a.a. - 437 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: TEEEKILYLTPEQEKDKSHFTDKETGQEHELIESMPLLEWFANNYKKFGATLEIVTDKSQEGSQFVKGFGGIGGILRYRVDFQGMEYQGGDDEFFDLDDY
Specifications
Accession Number NP_004721.1
Gene ID (Entrez) 2107
Name eukaryotic translation termination factor 1
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias D5S1995, ERF, ERF1, MGC111066, RF1, SUP45L1, TB3-1
Gene Symbol ETF1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer