Learn More
ETC-159, MedChemExpress
Product Code. 16480138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16480138
Brand: Medchem Express HY18988
MedChemExpress ETC-159 (ETC-1922159) is a potent, orally available PORCN inhibitor. ETC-159 inhibits β-catenin reporter activity with an IC50 of 2.9 nM.
ETC-159 (ETC-1922159) is a potent, orally available PORCN inhibitor. ETC-159 inhibits β-catenin reporter activity with an IC50 of 2.9 nM.
Specifications
Chemical Name or Material ETC-159
CAS 1638250-96-0
Color White
Purity Grade Notes Research
Molecular Formula C19H17N7O3
Quantity 5 mg
Synonym ETC-1922159
Solubility Information DMSO : 50 mg/mL (127.75 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=CC=CC=C2)N=N1)CN3C=NC4=C3C(N(C)C(N4C)=O)=O
Molecular Weight (g/mol) 391.38
Formula Weight 391.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.