Learn More
Scharlab ETANOL 96% PSS 25L TILLST. + FÖRSÄKRAN
Brand:  Scharlab ET0003025P
ETANOL 96% PSS 25L TILLST. + FÖRSÄKRAN
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 11304074
  • ONLINE EXCLUSIVE PRICE
    9124.50 NOK
    6916.89 NOK / 25 L
    Save 2207.61 NOK
    24% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ethanol 96% v/v, extra pure, Pharmpur, Ph Eur, BP
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Ethanol