Learn More
β-Estradiol 17-acetate, MedChemExpress
Product Code. 16438426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16438426 Supplier Medchem Express Supplier No. HYB0708
 Restricted Item
Estimated Shipment: 01-12-2026
MedChemExpress β-Estradiol 17-acetate is a metabolite of estradiol.
β-Estradiol 17-acetate is a metabolite of estradiol.
Specifications
Chemical Name or Material β-Estradiol 17-acetate
CAS 1743-60-8
Color White
Purity Grade Notes Research
Molecular Formula C20H26O3
Quantity 100 mg
Synonym 1,3,5(10)-Estratriene-3,17β-diol 17-acetate
Solubility Information DMSO : 100 mg/mL (318.05 mM; Need ultrasonic)
SMILES C[C@@]1([C@H]2OC(C)=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 314.42
Formula Weight 314.42
Percent Purity 95.64%
Grade Research
Physical Form Powder