Learn More
Cole-Parmer™ Essentials Digital Counter Pen
Product Code. 12038918 Shop All Cole-Parmer Products
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
12038918 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12038918 Supplier Cole-Parmer™ Supplier No. 2061030
  • ONLINE EXCLUSIVE PRICE
    244.00 NOK
    226.64 NOK / Pack of 1
    Save 17.36 NOK
    7% Off
View all promotions
Estimated Shipment: 08-12-2026
The Counter Pen is a combination of marker and digital counter, designed for the lab and plant. Use it to count cell growth, inventory parts and tally samples received. The counter beeps each time a mark is made. LCD simultaneously displays total and can be reset at the touch of a button.
  • Automatically beeps every time a touch is made
  • East to read LCD displays up to 99,999
Specifications
Display Type LCD
Description Digital counter pen
Material Durable ABS plastic
Diameter (Metric) 1.5 cm
Length (Metric) 15.2 cm
Weight (English) 1 oz.
Weight (Metric) 28.35 g
For Use With (Application) Microbiology, cell counting, tally counting, inventory parts counting
Includes Cap