Learn More
ESAM Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28739-1-AP-150UL
 View more versions of this product
Product Code. 16836235
  • 4836.10 NOK / 150µL
Description
The ESAM antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ESAM. This antibody recognizes human, mouse, rat antigen. The ESAM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ESAM
Polyclonal
Unconjugated
150 μL
90952
IgG
Western Blot, ELISA
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse