missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlotinib, MedChemExpress
Description
Erlotinib (CP-358774) is a directly acting EGFR tyrosine kinase inhibitor, with an IC50 of 2 nM for human EGFR. Erlotinib reduces EGFR autophosphorylation in intact tumor cells with an IC50 of 20 nM. Erlotinib is used for the treatment of non-small cell lung cancer.
Specifications
Specifications
| Chemical Name or Material | Erlotinib |
| CAS | 183321-74-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H23N3O4 |
| Quantity | 100 mg |
| Synonym | CP-358774 NSC 718781 OSI-774 |
| Solubility Information | DMSO : 3.94 mg/mL (10.01 mM; ultrasonic and warming and heat to 40°C) |
| SMILES | C#CC1=CC=CC(NC2=NC=NC3=C2C=C(C(OCCOC)=C3)OCCOC)=C1 |
| Molecular Weight (g/mol) | 393.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?