Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With DIN Thread
Product Code. 10833581
Change view
Click to view available options
Diameter (Metric) Outer:
64 mm
85 mm
105 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) Outer unitSize
10833581 105 mm Pack of 10
10290050 85 mm Pack of 10
10755442 64 mm Pack of 10
3 options
Product Code. 10833581
Brand: Lenz Laborglasintrumente 03025158
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With DIN thread (GL) and screw cap (red)
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™ (flask), PBT (cap)
Specifications
Capacity (Metric) 500 mL
Thread Size 32 GL
Diameter (Metric) Outer 105 mm
Quantity 10/Pk.